Learn More
Biomatik Corporation Human Pappalysin 2 (PAPPA2) CLIA Kit
Supplier:  Biomatik Corporation EKU09858
Encompass_Preferred
Human Pappalysin 2 (PAPPA2) CLIA Kit for detecting PAPPA2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9BXP8. Gene ID: 60676. Target Synonyms: PAPP-A2; PAPPE; PLAC3; Placenta-Specific 3; Pregnancy-associated plasma protein A2; Pregnancy-associated plasma protein E1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6632