Learn More
Biomatik Corporation Mouse Advanced Oxidation Protein Products (AOPP) ELISA Kit
Supplier:  Biomatik Corporation EKU10613
Encompass_Preferred
Mouse Advanced Oxidation Protein Products (AOPP) ELISA Kit for detecting AOPP in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 308.6-25,000ng/ml. Sensitivity: 104.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6716