Learn More
Biomatik Corporation High Sensitive Human Microtubule Associated Protein Tau (MAPt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10302
Encompass_Preferred
The High Sensitive Human Microtubule Associated Protein Tau (MAPt) ELISA Kit allows for quantitative determination of Microtubule Associated Protein Tau in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10636. Cross Activity: No significant cross-reactivity or interference between Microtubule Associated Protein Tau and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4512