Learn More
Biomatik Corporation Rat Troponin T (TNT) ELISA Kit
Supplier:  Biomatik Corporation EKU11492
Encompass_Preferred
Rat Troponin T (TNT) ELISA Kit for detecting TNT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Tn-T; TNNT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6935