Learn More
Biomatik Corporation Human/Mouse/Rat/Rabbit/Dog/Pig/Bovine/Horse Ubiquitin (Ub) ELISA Kit
Supplier:  Biomatik Corporation EKU11565
Encompass_Preferred
Human/Mouse/Rat/Rabbit/Dog/Pig/Bovine/Horse Ubiquitin (Ub) ELISA Kit for detecting Ub in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Pig (Sus scrofa; Porcine), Bovine (Bos taurus), Horse (Equus caballus; Equine). Detection Range: 4.94-400ng/ml. Sensitivity: 1.92ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P0CG47. Gene ID: 7314. Target Synonyms: Ubiquitin B; UbB; Ubiquitous Immunopoietic Polypeptide; Polyubiquitin B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6951