Learn More
Biomatik Corporation Cattle Xanthine Dehydrogenase (XDH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08231
Encompass_Preferred
The Cattle Xanthine Dehydrogenase (XDH) ELISA Kit allows for quantitative determination of Xanthine Dehydrogenase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80457. Cross Activity: No significant cross-reactivity or interference between Xanthine Dehydrogenase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4162