Learn More
Biomatik Corporation Monkey Interleukin-8, Il-8 Elisa Kit
Supplier:  Biomatik Corporation EKC36181
Encompass_Preferred
Monkey Interleukin-8, Il-8 Elisa Kit for detecting Interleukin 8 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P67813; CXCL8. Target Synonyms: CXCL8; IL8Interleukin-8; IL-8; C-X-C motif chemokine 8; Chemokine; C-X-C motif) ligand 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7565