Learn More
Biomatik Corporation Mouse C-Reactive Protein, Crp Elisa Kit
Supplier:  Biomatik Corporation EKC36670
Encompass_Preferred
Mouse C-Reactive Protein, Crp Elisa Kit for detecting C-Reactive Protein, Pentraxin-Related in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14847; Crp. Target Synonyms: Crp; Ptx1C-reactive protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7587