Learn More
Biomatik Corporation Pig Brain-derived neurotrophic factor(BDNF) ELISA kit
Supplier:  Biomatik Corporation EKC38112
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 9.375 ng/mL-600 ng/mL. Sensitivity: 2.35 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P14082. Target Synonyms: MGC34632, neurotrophin. Restriction: For Research Use Only.
Catalog No. 50-228-7630