Learn More
Biomatik Corporation Plant hormone abscisic acid, ABA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31457
Encompass_Preferred
Plant Hormone Abscisic Acid, Aba Elisa Kit for detecting Hormone Abscisic Acid, Aba in plant tissues. Species Reactivity: Plant. Detection Range: 0.156-10ug/ml. Sensitivity: 0.04ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-7463