Learn More
Biomatik Corporation Lipopolysaccharide (LPS) ELISA Kit
Supplier:  Biomatik Corporation EKU11047
Encompass_Preferred
Lipopolysaccharide (LPS) ELISA Kit for detecting LPS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 0.47-30ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: LOS; Lipoglycans; Lipooligosaccharide; Lipo-Oligosaccharide; Endotoxin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6846