Learn More
Biomatik Corporation Taurine (Tau) ELISA Kit
Supplier:  Biomatik Corporation EKU11459
Encompass_Preferred
Taurine (Tau) ELISA Kit for detecting Tau in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000ng/ml. Sensitivity: 4.39ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 2-Aminoethanesulfonic Acid; Tauric Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6930