Learn More
Biomatik Corporation Human Placental Alkaline Phosphatase, Plap Elisa Kit
Supplier:  Biomatik Corporation EKC35028
Encompass_Preferred
Human Placental Alkaline Phosphatase, Plap Elisa Kit for detecting Alkaline Phosphatase, Placental (Regan Isozyme) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 84-2000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05187; ALPP. Target Synonyms: ALPP; PLAP; Alkaline phosphatase; placental type; Alkaline phosphatase Regan isozyme; Placental alkaline phosphatase 1; PLAP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7517