Learn More
Biomatik Corporation Rat Mast Cell Tryptase,MCT ELISA Kit
Supplier:  Biomatik Corporation EKC39382
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P27435. Target Synonyms: TPS1, TPS2, TPSB1, mast cell alpha II tryptase|mast cell beta I tryptase|tryptase alpha II|tryptase beta 1|tryptase beta I|tryptase-I|tryptase-III. Restriction: For Research Use Only.
Catalog No. 50-228-7670