Learn More
Biomatik Corporation Human CPR (Cytochrome P450 Reductase) ELISA Kit
Supplier:  Biomatik Corporation EKF57580
Encompass_Preferred
Human CPR (Cytochrome P450 Reductase) ELISA Kit for detecting CPR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16435. Target Synonyms: CPR; CYPOR; Cytochrome P450 Reductase; P450R; EC 1.6.2.4; NADPH--cytochrome P450 reductase; NADPH-dependent cytochrome P450 reductase; P450; cytochrome oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7973