Learn More
Biomatik Corporation Human Family With Sequence Similarity 3, Member D (FAM3D) ELISA Kit
Supplier:  Biomatik Corporation EKU11814
Encompass_Preferred
Human Family With Sequence Similarity 3, Member D (FAM3D) ELISA Kit for detecting FAM3D in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96BQ1. Gene ID: 131177. Target Synonyms: EF7; OIT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6999