Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha (TNFa) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU11869
Encompass_Preferred
Mouse Tumor Necrosis Factor Alpha (TNFa) Wide-range ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06804. Gene ID: 21926. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7012