Learn More
Biomatik Corporation Human Collagen Type IV (COL4) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU11878
Encompass_Preferred
Human Collagen Type IV (COL4) High-sensitive ELISA Kit for detecting COL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02462. Target Synonyms: Type-IV Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7014