Learn More
Biomatik Corporation Human Endosialin (Cd248) Elisa Kit
Supplier:  Biomatik Corporation EKC33563
Encompass_Preferred
Human Endosialin (Cd248) Elisa Kit for detecting Cd248 Molecule, Endosialin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9HCU0; CD248. Target Synonyms: CD164 sialomucin like 1; CD164L1; CD248; CD248 antigen endosialin; CD248 molecule endosialin; CD248_HUMAN; Endosialin; TEM 1; TEM1; Tumor endothelial marker 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7402