Learn More
Biomatik Corporation Human Tartrate Resistant Acid Phosphatase (Trap) Elisa Kit
Supplier:  Biomatik Corporation EKC35710
Encompass_Preferred
Human Tartrate Resistant Acid Phosphatase (Trap) Elisa Kit for detecting Tartrate Resistant Acid Phosphatase(Trap) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30ng/ml. Sensitivity: 0.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7547