Learn More
Biomatik Corporation Human Cytokeratin 18-M30, Ck 18-M30 Elisa Kit
Supplier:  Biomatik Corporation EKC42319
Encompass_Preferred
Human Cytokeratin 18-M30, Ck 18-M30 Elisa Kit for detecting Cytokeratin 18-M30, Ck 18-M30 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000mIU/ml. Sensitivity: 3.9mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7866