Learn More
Biomatik Corporation Human Opi (Opiorphin) ELISA Kit
Supplier:  Biomatik Corporation EKF58246
Encompass_Preferred
Human Opi (Opiorphin) ELISA Kit for detecting Opi in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q99935. Target Synonyms: Opi; Opiorphin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8076