Learn More
Biomatik Corporation Rat Ceruloplasmin (CP) ELISA Kit
Supplier:  Biomatik Corporation EKL54173
Encompass_Preferred
Rat Ceruloplasmin (CP) ELISA Kit for detecting CP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.44ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 4h, 30min. Accession Number: P13635. Gene ID: 24268. Target Synonyms: CP2; CER; Caeruloplasmin; Ferroxidase; Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8581