Learn More
Biomatik Corporation Gas6 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52043
Encompass_Preferred
Gas6 (Human) Sandwich ELISA Kit for detecting Gas6 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14393. Gene ID: 2621. Target Synonyms: Growth arrest-specific protein 6; AXL receptor tyrosine kinase ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9171