Learn More
Biomatik Corporation ST2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52064
Encompass_Preferred
ST2 (Human) Sandwich ELISA Kit for detecting ST2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q01638. Gene ID: 9173. Target Synonyms: Interleukin-1 receptor-like 1; Protein ST2; DER4; T1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9179