Learn More
Biomatik Corporation Keratin 20 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50340
Encompass_Preferred
Keratin 20 Colorimetric Cell-Based ELISA Kit for detecting Keratin 20 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35900. Gene ID: 54474. Target Synonyms: Keratin type I cytoskeletal 20; Cytokeratin-20; CK-20; K20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0611