Learn More
Biomatik Corporation Pim-1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50480
Encompass_Preferred
Pim-1 Colorimetric Cell-Based ELISA Kit for detecting Pim-1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11309. Gene ID: 5292. Target Synonyms: PIM-1; Proto-oncogene serine/threonine-protein kinase pim-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0732