Learn More
Biomatik Corporation Rat Dickkopf Related Protein 1 (DKK1) ELISA Kit
Supplier:  Biomatik Corporation EKU03731
Encompass_Preferred
Rat Dickkopf Related Protein 1 (DKK1) ELISA Kit for detecting DKK1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z9J1. Gene ID: 293897. Target Synonyms: SK; Dickkopf Homolog 1 (Xenopus Laevis). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2307