Learn More
Biomatik Corporation Mouse Glypican 4 (GPC4) ELISA Kit
Supplier:  Biomatik Corporation EKU04547
Encompass_Preferred
Mouse Glypican 4 (GPC4) ELISA Kit for detecting GPC4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51655. Gene ID: 14735. Target Synonyms: K-Glypican; Glypican Proteoglycan 4; Secreted glypican-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2457