Learn More
Biomatik Corporation Human Klotho Beta (KLb) ELISA Kit
Supplier:  Biomatik Corporation EKU05513
Encompass_Preferred
Human Klotho Beta (KLb) ELISA Kit for detecting KLb in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86Z14. Gene ID: 152831. Target Synonyms: Klotho beta-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2599