Learn More
Biomatik Corporation Human Chemokine C-C-Motif Receptor 1 (CCR1) ELISA Kit
Supplier:  Biomatik Corporation EKU03097
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.235ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P32246. Target Synonyms: CD191; CMKBR1; HM145; MIP1aR; SCYAR1; RANTES-R; Macrophage inflammatory protein 1-alpha receptor; LD78 receptor. Restriction: For Research Use Only.
Catalog No. 50-229-2212