Learn More
Biomatik Corporation Human Chemokine (C-X-C Motif) Ligand 27 (CCL27) ELISA Kit
Supplier:  Biomatik Corporation EKU03526
Encompass_Preferred
Human Chemokine (C-X-C Motif) Ligand 27 (CCL27) ELISA Kit for detecting C-X-C Motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y4X3. Gene ID: 10850. Target Synonyms: CTACK; ILC; SCYA27; ALP; Skinkine; ESkine; PESKY; CTAK; Cutaneous T-Cell Attracting Chemokine; Small Inducible Cytokine Subfamily A Member 27; IL-11 R-alpha-locus chemokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2268