Learn More
Biomatik Corporation Mouse Pancreatic Elastase 1 (ELA1) ELISA Kit
Supplier:  Biomatik Corporation EKU03839
Encompass_Preferred
Mouse Pancreatic Elastase 1 (ELA1) ELISA Kit for detecting ELA1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91X79. Gene ID: 109901. Target Synonyms: PE; CELA1; Chymotrypsin-Like Elastase Family,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2334