Learn More
Biomatik Corporation Human Guanylate Cyclase 1 Beta 3 (GUCY1b3) ELISA Kit
Supplier:  Biomatik Corporation EKU04628
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q02153. Target Synonyms: GC-S-beta-1; GC-SB3; GUC1B3; GUCB3; GUCSB3; Soluble guanylate cyclase small subunit; Guanylate cyclase soluble subunit beta-3. Restriction: For Research Use Only.
Catalog No. 50-229-2472