Learn More
REPLIGEN CORPORATION ELISA KIT FOR MAB SELECT
SDP
Supplier:  REPLIGEN CORPORATION 93331
Encompass
Repligen Protein A ELISA for the detection of MabSelect SuRe. Used for downstream processes that utilize a MabSelect SuRe™ capture step. Repligen offers the only kit on the market that includes the SuRe™ ligand standard for the most accurate quantitation. Includes 3 sample prep methods, Dilute and Go (no boiling or dialysis), Boil & Boost (low conc. samples), and Original.
Catalog No. NC0027103