Learn More
Agdia Inc NPT II (neomycin phosphotransferase II) ELISA Kit
SDP
Supplier:  Agdia Inc PSP73000/0096
Encompass
The Agdia NPTII (neomycin phosphotransferase II) ELISA is intended for the detection or the absence of the NPTII marker gene protein expressed in various NPTII transformed crops. This kit is presented as a qualitative assay; however a certified reference standard (LST 73000) can be purchased for quantitative purposes. A quantitative user guide is available on our website or can be emailed to you upon request.
Catalog No. NC1639076