Learn More
BioVendor, LLC ELISA LEPTIN MOUSE/RAT (96 wells)
SDP
Supplier:  BioVendor, LLC RD291001200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements7 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range100–4000 pg/mlLimit of DetectionAnalytical Limit of Detection is calculated from the real leptin values in wells and is 30 pg/ml for mouse leptin and 50 pg/ml for rat leptin
Catalog No. 50-894-221