Learn More
Medchemexpress LLC Ellagic acid | 476-66-4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB01831G
Encompass_Preferred
Ellagic acid is a natural antioxidant that acts as a potent and ATP-competitive inhibitor of CK2 and SHP2, with an IC50 of 40 nM and a Ki of 20 nM. This compound also blocks other kinases such as LYN, PKA, SYK, GSK3, FGR, and CK1.
  • Acts as a natural antioxidant
  • Functions as a potent and ATP-competitive inhibitor of CK2 and SHP2
  • Blocks various other kinases including LYN, PKA, SYK, GSK3, FGR, and CK1
  • Exhibits inhibitory activities against Karpas299, SUDHL1, SR786, and FE-PD cell lines
  • Shows cytotoxic effects on MCF-7 cells after radiation treatment
  • Induces cell apoptosis in MCF-7 cells when combined with irradiation
  • Decreases MDA brain content and reduces brain TNF-α levels in rats
  • Increases brain contents of 5-HT, dopamine, and norepinephrine in rats
  • Reduces histopathological changes induced by Doxorubicin in rats
  • Supported by over 20 cited publications
Catalog No. 50-004-05758