Learn More
Medchemexpress LLC Ellagic acid 3-O-α-L-rhamnopyranoside | 1422265-57-3 | 98.0% | 448.33 | 10 MG

Supplier: Medchemexpress LLC 10MGHYN12350
Ellagic acid 3-O-α-L-rhamnopyranoside is an ellagic acid derivative that can be isolated from Folium Turpiniae. It is intended for research use only and has not been fully validated for medical applications.
- Isolated from Folium Turpiniae
- Appears as an off-white to light yellow solid
- Stable under recommended storage conditions
- Recommended storage for powder: -20°C for 3 years
- Recommended storage for in solvent: -80°C for 6 months, or -20°C for 1 month
- Not classified as a hazardous substance or mixture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.