Learn More
Medchemexpress LLC Ellagic acid 3-O-α-L-rhamnopyranoside | 1422265-57-3 | 98.0% | 448.33 | 1 MG

Supplier: Medchemexpress LLC 1MGHYN12350
Ellagic acid 3-O-α-L-rhamnopyranoside is an ellagic acid derivative that can be isolated from Folium Turpiniae. It is intended for research use only and not sold to patients.
- An ellagic acid derivative.
- Isolated from Folium Turpiniae.
- For research use only.
- Appearance: Solid.
- Color: Off-white to light yellow.
- Shipping: Room temperature in continental US.
- Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, or -20°C for 1 month.
- Documents available: Data Sheet, SDS, COA, HNMR, LCMS, Handling Instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.