Learn More
Medchemexpress LLC Ellagic acid (dihydrate) | 133039-73-3 | 98.4% | 100 MG

Supplier: Medchemexpress LLC HYB0183B100MG
Ellagic acid (dihydrate) is a biochemical reagent for life science related research, usable as a biological material or organic compound. It is intended for research use only and is not sold to patients.
- Purity of 98.42%.
- Solid appearance.
- Light yellow to brown color.
- Molecular weight: 338.22.
- Molecular formula: C14H10O10.
- Store at 4°C, sealed, away from moisture.
- When in solvent, store at -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.