Learn More
Chemglass Life Sciences Ellipso-Spoon, 15 x 35 x 210mm
SDP
Supplier:  Chemglass Life Sciences CG198702
Encompass_Preferred
Ellipso-Spoon, 15 x 35 x210mm. The specialty curved shape permits contact with all corners of bottles, flasks and containers. Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula end and is made of polished stainless steel. Has a 5mm diameter handle.
Catalog No. 50-872-451