Learn More
Bachem 3,4-Dihydro-2H-pyran-2-ylmethoxymethyl resin (200-400 mesh, 0.7-1.0 mmol/g)
SDP
Supplier:  Bachem 4028707.0005
Encompass_Preferred
3,4-Dihydro-2H-pyran-2-ylmethoxymethyl resin (200-400 mesh, 0.7-1.0 mmol/g), 5g. Ellman's dihydropyran resin. Ellman's dihydropyran resin allows coupling primary and secondary as well as functionalized alcohols as acetal with high loading levels. The attachment functionality was stable to both strongly basic and nucleophilic reagents. Ellman’s resin has been used within the synthesis of ketones from esters and biphenyltetrazole derivatives. Release of the product alcohol from the support is carried out by treating with 95% TFA / 5% water, TFA / CH2Cl2 / EtOH or pyridinium p-toluenesulfonate / BuOH / 1,2-dichloroethane.
Catalog No. 50-194-7377