Learn More
TARGETMOL CHEMICALS INC ELN318463 RACEMATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11178L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ELN318463 racemate (ELN 318463 racemate) is the racemate of ELN318463 which is an amyloid precursor protein (APP) selective (gamma)-secretase inhibitor. purity: 99%
Catalog No. 50-258-1556