Learn More
Medchemexpress LLC ELN484228 10mg | 312-63-0 | 10 MG

Supplier: Medchemexpress LLC HY11503810MG
ELN484228 is a small-molecule blocker of α-synuclein used in Parkinson's disease research. It is provided as a high-purity solid suitable for in vitro and in vivo pharmacology studies and is soluble in DMSO with compatible in vivo formulations.
- High purity (99.5%).
- Molecular weight 251.28 g/mol.
- Chemical formula C12H10FNO2S.
- Physical form solid powder.
- Solubility ≥50 mg/mL in DMSO; in vivo formulations documented.
- Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
- Available in small milligram pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.