Learn More
Medchemexpress LLC Eloralintide (sodium) | 99.7% | 4526.10 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP10798A1MG
Encompass_Preferred
Eloralintide sodium is an Amylin Receptor agonist that selectively activates AMY1R and AMY3R in cells expressing human or rat AMY1R, AMY3R or CTR. It activates AMYRs and CTR in rats, leading to a sustained decrease in plasma Ca2+ levels. Eloralintide sodium reduces appetite and body weight in both lean and obese rats and can be used for research related to obesity.
  • Selectively activates AMY1R and AMY3R
  • Activates AMYRs and CTR in rats
  • Leads to a sustained decrease in plasma Ca2+ levels
  • Reduces appetite and body weight in lean and obese rats
  • Useful for obesity research
Catalog No. 50-004-22951