Learn More
Medchemexpress LLC Elovl1-IN-2 | 2761063-79-8 | 99.8% | C18H15FN2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14527050MG
Encompass_Preferred
ELOVL1-IN-2 is an inhibitor of elongation of very long chain fatty acid 1 (ELOVL1) enzyme. It exhibits weak ELOVL1 inhibition with an IC50 of 21 μM. In a primary cellular assay using HEK293 C26 cells, it shows moderate potency with an IC50 of 6.7 μM.
  • ELOVL1 enzyme inhibitor
  • Weak ELOVL1 inhibition (IC50 = 21 μM)
  • Moderate potency in HEK293 C26 cellular assay (IC50 = 6.7 μM)
Catalog No. 50-003-56214