Learn More
Medchemexpress LLC Elraglusib | 1034895-42-5 | 99.1% | 404.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11391450MG
Encompass_Preferred
9-ING-41 (Elraglusib) is an ATP-competitive and selective inhibitor of glycogen synthase kinase-3β (GSK-3β), characterized by its maleimide-based structure and an IC50 of 0.71 μM. This compound plays a role in inducing cell cycle arrest, autophagy, and apoptosis in cancer cells, thereby exhibiting anticancer activity. It also holds potential for augmenting the antitumor effects of various chemotherapeutic agents.
  • Leads to cell cycle arrest, autophagy, and apoptosis in cancer cells.
  • Possesses anticancer activity.
  • Potential for enhancing the antitumor effects of chemotherapeutic agents.
  • Exhibits single-agent antitumor activity in a mouse model of MCL.
Catalog No. 50-004-37837