Learn More
Cayman Chemical ElsIglutIdeacetate 10mg
SDP
Supplier:  Cayman Chemical 4127310 MG
Encompass_Preferred
A peptide; increases jejunum and ileum crypt depth and villus height without disrupting gut microbiota diversity in rats at 0.9 mg/kg four days per week; decreases lapatinib-induced diarrhea severity and incidence in rats; prevents lapatinib-induced loss of body weight in rats
Catalog No. 50-294-4947