Learn More
Selleck Chemical LLC Eltanexor (KPT-8602) 2mg 1642300-52-4 ONO-7706ATG-016
SDP
Supplier:  Selleck Chemical LLC S83972MG
Encompass_Preferred
Eltanexor (KPT-8602, ONO-7706,ATG-016) is a second-generation, orally bioavailable XPO1 (also known as CRM1) inhibitor with IC50 values of 20?211?nM in 10 AML lines after 3 days exposure. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6024